Learn More
Thermo Scientific™ Oil-Free Vacuum Pumps for Vacuum Concentrators
Eliminate oil mist vapors with these quiet-running, chemical resistant, oil-free pumps, designed for a wide variety of laboratory applications.
10050.00 €
Specifications
Type | Oil-Free Vacuum Pump |
---|---|
For Use With (Equipment) | Vacuum Concentrator |
Description
Eliminate oil mist vapors with Thermo Scientific™ Oil-Free Vacuum Pumps for Vacuum Concentrators, designed for a wide variety of laboratory uses. These quiet-running pumps are highly chemical resistant, and do not require oil to operate.
- Small footprint
- Four-headed design with chemically resistant solid ETFE and PFA heads
- Easily connects to any Thermo Scientific™ SpeedVac™ system
- Recommended for use with acidic environments such as HCl, TFA, Acetic and Formic Acids as well as organic solvents
- Comes complete with ANT100 Post Trap for adding trapping and neutralizing solutions
Specifications
Oil-Free Vacuum Pump | |
Vacuum Concentrator |