missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oil Red O, Biological Stain

Product Code. 15244664
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1320-06-5
Molecular Formula C26H24N4O
Molecular Weight (g/mol) 408.51
MDL Number MFCD00003898
InChI Key HJAYEODIXUYVIC-LTFAAIOASA-N
Synonym C.I. 26125, Solvent Red 27
PubChem CID 6046885
SMILES CC1=CC=C(C)C(=C1)N=NC1=CC(C)=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C=C1C

Specifications

IUPAC Name (1Z)-1-[[4-[(2,5-dimethylphenyl)diazenyl]-2,5-dimethylphenyl]hydrazinylidene]naphthalen-2-one
Formula Weight 408.5
Quantity 25 g
Chemical Name or Material Oil Red O

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'