Learn More
Justrite™ Oily Waste Can Foot Operated Cover & Soundgard
Foot operated for convenience and ease of use.
Brand:  Justrite™ 09308
Product Code. 12903565
  • 214.00 € / Each
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Rags and cloths soaked with solvents, thinners, linseed oil, combustible adhesives and other flammable liquids present a serious fire risk when improperly discarded. Lid opens to no more than 60 degrees and stays closed when not in use, isolating contents from fire sources and limiting oxygen to virtually eliminate the risk of spontaneous combustion.
Specifications
Specifications
13.94 in.
18.25 in.