missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Antagonist of 5-HT2A and dopamine D2 receptors (Ki values are 4 and 11 nM respectively). Also displays affinity for a range of other receptors including D1 and D4, 5-HT2C, α1, H1 and M1-4 receptors (Ki values range between 1.9 and 31 nM) . Atypical antipsychotic. Displays anticholinergic properties. Also a highly potent activator of hM4Di DREADDs (EC50 values are 5 nM in vitro and 7 nM in vivo).

Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 312.43 |
Percent Purity | >99% |
Chemical Name or Material | Olanzapine |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Olanzapine, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction