missing translation for 'onlineSavingsMsg'
Learn More

Oleic acid, 99%, Thermo Scientific Chemicals

Product Code. 11388307
Click to view available options
Quantity:
5 g
25 g
100 g

Oleic acid is used as an emulsifying agent for facial creams and lotions, as an accelerator in the rubber industry, as an emollient, as a solubilizing agent in aerosol products and as a soldering flux in stained glass work for joining lead came. It is used in the preparation of oleates and lotions. It acts as intermediate for drilling and fracking fluids in the oil and gas industry. It finds application as an excipient in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-80-1
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.47
MDL Number MFCD00064242
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
PubChem CID 445639
ChEBI CHEBI:16196
IUPAC Name (Z)-octadec-9-enoic acid
SMILES CCCCCCCC\C=C\CCCCCCCC(O)=O

Specifications

Melting Point 13°C to 14°C
Color Colorless to Yellow
Density 0.887 g/mL
Boiling Point 286°C (100 mmHg)
Refractive Index 1.461
Quantity 25 g
Beilstein 1726542
Sensitivity Air Sensitive
Merck Index 146828
Solubility Information Miscible with ethanol,ether,acetone,chloroform,dimethyl formamide and dimethyl sulfoxide.
Formula Weight 282.47
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Oleic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.