missing translation for 'onlineSavingsMsg'
Learn More

Oleic acid, Honeywell™

Product Code. 15617040
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 112-80-1
Density 0.89g/mL at 25°C
Flash Point <113°C (>235.4°F)
Molecular Formula C18H34O2
Linear Formula CH3(CH2)7CH=CH(CH2)7COOH
MDL Number MFCD00064242
Quantity 1 L
UN Number NONH for all modes of transport
Beilstein 1726542
Synonym oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
SMILES CCCCCCCC\C=C\CCCCCCCC(O)=O
IUPAC Name (9E)-octadec-9-enoic acid
Molecular Weight (g/mol) 282.47
PubChem CID 445639
ChEBI CHEBI:16196
Formula Weight 282.46g/mol
Vapor Pressure 1 mmHg (176°C)
Chemical Name or Material Oleic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.