missing translation for 'onlineSavingsMsg'
Learn More

Oleic acid, sodium salt, 65-90% oleic C18, Thermo Scientific Chemicals

Product Code. 10625862
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 143-19-1
Melting Point 232°C to 235°C
Color White to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C18H33NaO2
Linear Formula CH3(CH2)7CH=CH(CH2)7CO2Na
MDL Number MFCD00004438
Quantity 1 g
Beilstein 02,I,198
Merck Index 15, 6921
Synonym sodium oleic acid
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
InChI Key BCKXLBQYZLBQEK-KVVVOXFISA-M
SMILES CCCCCCCCC=CCCCCCCCC(=O)[O-].[Na+]
IUPAC Name sodium;(Z)-octadec-9-enoate
Molecular Weight (g/mol) 304.44
PubChem CID 23665730
ChEBI CHEBI:81860
Formula Weight 304.44
Percent Purity 65-90% oleic C18
Physical Form Powder
Chemical Name or Material Oleic acid, sodium salt, 65-90% oleic C18
Show More Show Less
Product Identifier
  • Oleic acid, sodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'