Learn More
Oleyl alcohol, tech. 80-85%, Thermo Scientific Chemicals
27.54 € - 64.12 €
Chemical Identifiers
CAS | 143-28-2 |
---|---|
Molecular Formula | C18H36O |
Molecular Weight (g/mol) | 268.49 |
MDL Number | MFCD00002993 |
InChI Key | ALSTYHKOOCGGFT-KTKRTIGZSA-N |
Synonym | oleyl alcohol, cis-9-octadecen-1-ol, z-octadec-9-en-1-ol, ocenol, conditioner 1, dermaffine, lancol, novol, oceol, oleol |
PubChem CID | 5284499 |
ChEBI | CHEBI:73504 |
IUPAC Name | (9Z)-octadec-9-en-1-ol |
SMILES | CCCCCCCC\C=C/CCCCCCCCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11449156
|
Thermo Scientific Alfa Aesar
A18018.22 |
100 g |
27.54 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459156
|
Thermo Scientific Alfa Aesar
A18018.36 |
500 g |
64.12 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Oleyl alcohol is used in softening and lubrication of textile fabrics, and in the production of carbon paper, stencil paper, and printing ink. It finds application in cosmetic products viz skin creams and lotions as a thickner, hair conditioners and hair coating shampoos. It is utilized as an antifoaming agent and cutting lubricant, as the precursor for the preparation of its sulfuric ester derivatives, which are used in detergents and wetting agents. It plays a vital role in various formulations for drug delivery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsOleyl alcohol is used in softening and lubrication of textile fabrics, and in the production of carbon paper, stencil paper, and printing ink. It finds application in cosmetic products viz skin creams and lotions as a thickner, hair conditioners and hair coating shampoos. It is utilized as an antifoaming agent and cutting lubricant, as the precursor for the preparation of its sulfuric ester derivatives, which are used in detergents and wetting agents. It plays a vital role in various formulations for drug delivery.
Solubility
Miscible with alcohol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.
Notes
Incompatible with oxidizing agents and strong acids.
Chemical Identifiers
143-28-2 | |
268.49 | |
ALSTYHKOOCGGFT-KTKRTIGZSA-N | |
5284499 | |
(9Z)-octadec-9-en-1-ol |
C18H36O | |
MFCD00002993 | |
oleyl alcohol, cis-9-octadecen-1-ol, z-octadec-9-en-1-ol, ocenol, conditioner 1, dermaffine, lancol, novol, oceol, oleol | |
CHEBI:73504 | |
CCCCCCCC\C=C/CCCCCCCCO |
Specifications
143-28-2 | |
0.849 | |
180°C (356°F) | |
C18H36O | |
MFCD00002993 | |
oleyl alcohol, cis-9-octadecen-1-ol, z-octadec-9-en-1-ol, ocenol, conditioner 1, dermaffine, lancol, novol, oceol, oleol | |
ALSTYHKOOCGGFT-KTKRTIGZSA-N | |
(9Z)-octadec-9-en-1-ol | |
5284499 | |
268.48 | |
Liquid |
∽6°C | |
206°C to 207°C (13 mm) | |
0.85 | |
1.46 | |
1723962 | |
Miscible with alcohol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water. | |
CCCCCCCC\C=C/CCCCCCCCO | |
268.49 | |
CHEBI:73504 | |
Technical | |
Oleyl alcohol |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'einecsNumber' : 205-597-3
missing translation for 'rtecsNumber' : RG4120000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.