missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oleylamine, approximate C18-content 80-90%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
100 mL
1 L
5 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Yellow |
Density | 0.8100g/mL |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Linear Formula | CH3(CH2)7CH=CH(CH2)8NH2 |
Refractive Index | 1.4585 to 1.4625 (20°C, 589nm) |
Quantity | 5 mL |
IUPAC Name | (E)-octadec-9-en-1-amine |
Formula Weight | 267.5 |
Percent Purity | ≥96.0% (primary amine) |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Oleylamine, approximate C18-content 80-90%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction