missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oleylamine, approximate C18-content 80-90%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
100 mL
1 L
5 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Yellow |
Density | 0.8100g/mL |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Linear Formula | CH3(CH2)7CH=CH(CH2)8NH2 |
Refractive Index | 1.4585 to 1.4625 (20°C, 589nm) |
Quantity | 1 L |
IUPAC Name | (E)-octadec-9-en-1-amine |
Formula Weight | 267.5 |
Percent Purity | ≥96.0% (primary amine) |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0454474 | Certificate of Analysis | 25/08/2023 | 129540051,129540050,129540010,129541000 |
A0403653 | Certificate of Analysis | 15/12/2022 | 129540051,129540050,129540010,129541000 |
A0400448 | Certificate of Analysis | 15/12/2022 | 129540051,129540050,129540010,129541000 |
A0394811 | Certificate of Analysis | 15/12/2022 | 129540051,129540050,129540010,129541000 |
A0392078 | Certificate of Analysis | 15/12/2022 | 129540051,129540050,129540010,129541000 |
Safety and Handling




Product Identifier
- Oleylamine
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P273-Avoid release to the environment.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Oleylamine, approximate C18-content 80-90%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction