Learn More
Thermo Scientific™ Oligo(dT)18 Primer
Thermo Scientific Oligo(dT)18 Primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends.
Brand: Thermo Scientific™ SO131
73.63 EUR valid until 2025-02-28
Use promo code "24154" to get your promotional price.
Description
Thermo Scientific Oligo(dT)18 Primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends. The primer is supplied as a ready-to-use, 20X concentrated aqueous solution.
Applications
• cDNA Synthesis
• RT-qPCR
• RT-PCR
Related products
Anchored Oligo dT (Cat. No. AB1247)
Random Hexamer Primer (Cat. No. SO142)

Specifications
Primer | |
18-mer | |
0.5 μg/μL | |
60 μL | |
Liquid |
• Oligo(dT)18 Primer (60 μL at 0.5 μg/μL) Store at –20°C. |
|
Dry Ice | |
Oligo dT | |
cDNA Synthesis |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.