Learn More
Olopatadine hydrochloride, MedChemExpress
MedChemExpress Olopatadine hydrochloride (ALO4943A) is a histamine blocker used to treat allergic conjunctivitis.
Brand: Medchem Express HY-B0426A
This item is not returnable.
View return policy
Description
Olopatadine hydrochloride (ALO4943A) is a histamine blocker used to treat allergic conjunctivitis.Specifications
Olopatadine hydrochloride | |
White | |
C21H24ClNO3 | |
ALO4943A KW4679 | |
O=C(O)CC1=CC=C(C/2=C1)OCC3=CC=CC=C3C2=C\CCN(C)C.Cl | |
373.87 | |
Research | |
COVID-19-immunoregulation |
140462-76-6 | |
Research | |
10 mg | |
DMSO : 50 mg/mL (133.74 mM; Need ultrasonic) ∣H2O : 6.67 mg/mL (17.84 mM; Need ultrasonic) | |
373.87 | |
99.97% | |
Solid |