Learn More
Thermo Scientific™ Olympus™ 1.25X Objective, PlanApo N, 0.04 NA
Expand the functionality of your Thermo Scientific™ high-content instrument
Brand:  Thermo Scientific™ N010125
Product Code. 17236583
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Expand the functionality of your Thermo Scientific™ high-content instrument
Add a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific™ ArrayScan™ HCA Readers and the Thermo Scientific™ CellInsight™ NXT High Content Screening (HCS) Platform.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Objective Lens
Olympus™ 1.25X Objective, PlanApo N (0.04 NA)
Dry
1 each
Olympus
ArrayScan HCA Readers
1.25x (0.04NA)