Learn More
Invitrogen™ Olympus™ 20X Objective, fluorite, 0.45NA/6.6–7.8WD, correction collar (0–2.0 mm)
This fluorite (semi-apochromat) objective is equipped with an adjustable collar that affords flexibility in imaging through vessels with a thickness from 0 (no coverslip) to 2.0 mm. With four-color correction for spherical and chromatic aberrations, it delivers exceptional image quality.
Brand:  Invitrogen™ AMEP4763EO
Product Code. 16664245
  • 1862.00 € / Each
Explore more special offers
Description
Show More
Specifications
Specifications
Objective Lens
EVOS™
Fluorite with correction collar
1 each
Olympus
Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates
20X
Room Temperature
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Olympus™ 20X Objective, fluorite, 0.45NA/6.6–7.8WD, correction collar (0–2.0 mm) > Objective Lens; 20X; Fluorite; 0.45NA/6.6–7.8WD; Correction collar (0–2.0 mm)