Learn More
Thermo Scientific™ Olympus™ 2X Objective, PlanApo, 0.08 NA
Expand the functionality of your Thermo Scientific™ high-content instrument
Brand: Thermo Scientific™ NX1002X
This item is not returnable.
View return policy
Description
Expand the functionality of your Thermo Scientific™ high-content instrument
Add a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific™ ArrayScan™ HCA Readers and the Thermo Scientific™ CellInsight™ NXT High Content Screening (HCS) Platform.Specifications
Objective Lens | |
Olympus™ 2X Objective, PlanApo (0.08 NA) | |
Dry | |
1 each |
Olympus | |
CellInsight CX5 HCS Platform | |
2x (0.08NA) |