missing translation for 'onlineSavingsMsg'
Learn More

Omadacycline hydrochloride, MedChemExpress

Product Code. p-200100084
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496218

Brand: Medchem Express HY14865C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Omadacycline (PTK 0796) hydrochloride, a first-in-class orally active aminomethylcycline antibacterial, is a member of the tetracycline class of antibiotics.

Omadacycline hydrochloride acts through the inhibition of bacterial protein synthesis by binding to the 30S ribosomal subunit. Omadacycline hydrochloride possesses broad-spectrum antibacterial activity against aerobic and anaerobic Gram-positive and Gram-negative bacteria, as well as atypical bacteria. Omadacycline hydrochloride can be used for the research of acute bacterial skin and skin-structure infections, community-acquired pneumonia, and urinary tract infections.

Specifications

Chemical Name or Material Omadacycline hydrochloride
CAS 1196800-39-1
Color Dark Brown
Purity Grade Notes Research
Molecular Formula C29H40N4O7.HCl
Quantity 10 mg
Synonym PTK0796 hydrochloride Amadacycline hydrochloride
Solubility Information DMSO : 105 mg/mL (177.03 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (84.30 mM)
SMILES O=C(C1=C(O)[C@@H](N(C)C)[C@@](C[C@@]2([H])C(C(C3=C(O)C(CNCC(C)(C)C)=CC(N(C)C)=C3C2)=O)=C4O)([H])[C@@]4(O)C1=O)N.Cl
Molecular Weight (g/mol) 593.11
Formula Weight 593.11
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.