Learn More
Omadacycline tosylate, MedChemExpress
MedChemExpress Omadacycline (PTK 0796) tosylate, a first-in-class orally active aminomethylcycline antibacterial, is a member of the tetracycline class of antibiotics.
Brand: Medchem Express HY-14865B
This item is not returnable.
View return policy
Description
Omadacycline tosylate acts through the inhibition of bacterial protein synthesis by binding to the 30S ribosomal subunit. Omadacycline tosylate possesses broad-spectrum antibacterial activity against aerobic and anaerobic Gram-positive and Gram-negative bacteria, as well as atypical bacteria. Omadacycline tosylate can be used for the research of acute bacterial skin and skin-structure infections, community-acquired pneumonia, and urinary tract infections.Specifications
Omadacycline tosylate | |
Yellow | |
C36H48N4O10S | |
PTK 0796 tosylateAmadacycline tosylate | |
O=C(C1=C(O)[C@@H](N(C)C)[C@@](C[C@@]2([H])C(C(C3=C(O)C(CNCC(C)(C)C)=CC(N(C)C)=C3C2)=O)=C4O)([H])[C@@]4(O)C1=O)N.OS(=O)(C5=CC=C(C)C=C5)=O | |
728.85 | |
Research | |
COVID-19-immunoregulation |
1075240-43-5 | |
Research | |
50 mg | |
H2O : 12.5 mg/mL (17.15 mM; Need ultrasonic) | |
728.85 | |
99.37% | |
Powder |