Learn More
Diba Omnifit™ BenchMark™ Microbore Chromatography Columns
Optimized for aqueous systems and compatible with solvents used in common liquid chromatography applications like protein purification
Brand:  Diba 006BCC-03-02-FF
Encompass_Preferred
 View more versions of this product
Product Code. 15815254
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Includes: Ready-assembled column consisting of glass tube and two fixed endpieces preassembled with 2μm SS frits.
Description
Description
  • Economical, nonadjustable columns for low pressure or gravity feed
  • 3mm diameter and a variety of lengths
  • Compatible with a variety of bulk media
Not resistant to acetone, ketones, chlorinated hydrocarbons, aliphatic esters, phenol, >10% NaOH, >10% HCl, >5% acetic acid, or strong mineral acid.
Specifications
Specifications
3 mm
25 μm
25 mm