Learn More
Diba Omnifit™ BenchMark™ Replacement O-rings for Column Assemblies
Choose from FKM or FFKM O-rings
Brand:  Diba 006OR-35-V
Encompass_Preferred
 View more versions of this product
Product Code. 15835574
  • POA / Pack of 6
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
Use for quick-disconnect fittings, valves, pumps, and other fluid handling products.
Specifications
Specifications
O-ring
1.37 in.
Chromatography
102 mm
Black
35 mm
Omnifit Benchmark Chromatography Columns
FKM