missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diba Omnifit™ Chromatography Column Replacement Endpieces
No need to worry about losing an endpiece
Brand: Diba 006BFE-03-SS
This item is not returnable.
View return policy
Description
The endpieces make the primary seal to glass microbore columns.Specifications
Endpiece | |
0.51 cm | |
PTFE | |
3 mm | |
Columns | |
BenchMark Columns |
Fixed Length | |
3.81 cm | |
0.11 in. | |
0.2 μm | |
4/Pk. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction