Learn More
Diba Omnifit™ EZ Chromatography Columns with Two Fixed Endpieces
Omnifit™ EZ Chromatography Columns with Two Fixed Endpieces feature PEEK™ column threads which offer outstanding durability
Brand: Diba 006EZ-15-33-FF
This item is not returnable.
View return policy
Description
- Precision borosilicate glass columns provide inertness, visibility, and performance
- Superior frit placement adds bed support and even distribution of sample flow
- Reusable columns let you pack your own media specific to your application
Specifications
4 to 40°C | |
Liquid Chromatography Column | |
10 and 30 μm | |
330 mm | |
40°C |
15 mm | |
300 psi | |
53 mL | |
Borosilicate Glass, PTFE, Polyethylene, FKM/FPM | |
Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF Polypropylene fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits), and a Click-N-Seal™ 10-32 PEEK conical port fitting. |