Learn More
Diba Omnifit™ Packing Sleeves for EZ and EZ SolventPlus™ Columns
Use Omnifit™ Packing Sleeves for EZ and EZ SolventPlus™ Columns to join two columns for filling and packing
Brand:  Diba 006EPS-06
Encompass_Preferred
 View more versions of this product
Product Code. 15825274
  • 67.18 € / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Omnifit™ EZ and EZ SolventPlus™ Columns
6.6 mm
Liquid Chromatography Column