Learn More
Diba Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Columns
Mix and match your Omnifit™ Replacement Glass Tubes for EZ and EZ SolventPlus™ Column lengths while using your existing endpieces for ultimate flexibility
Brand: Diba 006ERG-25-25
This item is not returnable.
View return policy
Description
Use a different length tube with your existing endpieces to meet your specific requirements.Specifications
Omnifit™ EZ and EZ SolventPlus™ Columns | |
Liquid Chromatography Column | |
Glass |
25 mm | |
250 mm |