missing translation for 'onlineSavingsMsg'
Learn More

B Braun™ Omnifix™ Solo 3-Piece Syringes

Features smooth plunger action and double-seal stopper. B Braun™ Omnifix™ Solo 3-Piece Syringes are designed for consistent performance.

94.00 €

Specifications

Certifications/Compliance EN ISO 7886-1, ISO 10993 resp. DIN EN 30993
Material Polyisoprene Rubber (Tip), Polypropylene (Barrel), Polypropylene (Plunger)
Color Natural
Description High contrast black graduation
For Use With (Application) For slow aspiration and injection
View More Specs

Products 1
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
10179411
View Documents
B Braun™
4617100V
10 mL
94.00 €
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • High visibility; transparent barrel with permanent bold graduations
  • Secure backstop to prevent accidental plunger withdrawal
  • Double seal stopper prevents medication leakage
  • Large, stable finger grips
  • Components do not contain DEHP, PVC, or other rubber tree latex
  • Disposable
  • Piston with double sealing ring provides slow aspiration and injection
Specifications

Specifications

EN ISO 7886-1, ISO 10993 resp. DIN EN 30993
Natural
For slow aspiration and injection
Centric Luer-lock 3-part Syringe
Centric Luer Lock
Sterile
Polyisoprene Rubber (Tip), Polypropylene (Barrel), Polypropylene (Plunger)
High contrast black graduation
Yes
Single-use
Yes
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 5 Years

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.