Learn More
Oncomine™ Research Premium Edition License
Oncomine™ Research Premium Edition License
Brand: Invitrogen A18578
This item is not returnable.
View return policy
Description
Oncomine™ Research Premium Edition is a subscription-based software tool for academic researchers that provides additional advanced features and analyses over Oncomine™ Research Edition (a free edition available at www.oncomine.org). Oncomine™ Research Edition is a powerful web application that integrates and unifies high-throughput cancer profiling data so that target expression across a large number of cancer types and experiments can be accessed online, in seconds. Oncomine™ Research Edition includes basic analysis types such as cancer vs. normal, multi-cancer, and co-expression. It features gene and concept summaries, outlier analysis, meta-analysis, and meta-cancer outlier profile analysis (COPA). In addition, Oncomine™ Research Edition has an Oncomine™ support community site where users can share their experiences using this software tool.Oncomine™ Research Premium Edition is a subscription version that allows users with advanced research needs to take advantage of additional features and analyses. These features include both basic and advanced analyses such as cancer subtype, clinical outcome, patient treatment response, perturbation analyses, and drug sensitivity analyses. These analyses provide valuable insight into biology, regulation, pathways, drug response, and patient populations. In addition, the Premium Edition allows users to search for multiple genes at a time, export to Excel, PowerPoint, and SVG to share results, upload custom concepts, and access enhanced support, as well as search the Oncomine™ database for enrichment of user-defined gene signatures.
Oncomine™ Research Premium Edition features:
Learn more about Oncomine™ Research Premium Edition.
Oncomine™ Research Premium Edition can be licensed for various user numbers, up to an institutional license with unlimited number of registered users. You must be a registered user of Oncomine™ Research Edition to upgrade.
Note: This product is for academic and nonprofit cancer researchers only. If you are a commercial/for-profit cancer researcher, please contact one of our knowledgable sales specialists at compendiasales@lifetech.com.
How to Order
If you are not already a registered user of the free version of the software, please visit www.oncomine.org to register for Oncomine™ Research Edition. You will receive an email with your user name and temporary password, which you can then use to log in a get started using the free edition. Once this is done, or if you are already a registered use of the free edition, simply order this product as you would any standard product. During checkout you will be asked to provide information about each user of the software. Within 1 business day your account will be upgraded to the Premium edition and you will see the added features of the Premium edition the next time you log in.
Specifications
Oncomine™ | |
1 year, 2 users |
For Research Use Only. Not for use in diagnostic procedures.