missing translation for 'onlineSavingsMsg'
Learn More
Ondansetron hydrochloride dihydrate, 98%, Thermo Scientific Chemicals Product Code.: 15688715

Ondansetron hydrochloride dihydrate, 98%, Thermo Scientific Chemicals

Product Code. 15688715
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15688715

Brand: Thermo Scientific Acros 461160050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103639-04-9
Molecular Formula HCl·2H2O
Molecular Weight (g/mol) 365.85
InChI Key BGGIFKYQGVGSIW-UHFFFAOYSA-N
Synonym 1,2,3,9-tetrahydro-9-methyl-3-2-methyl-1h-imidazol-1-yl methyl-4h-carbazol-4-one hydrochloride; gr 38032f; ondansetron hydrochloride
PubChem CID 71317260
IUPAC Name chlorine;9-methyl-3-[(2-methylimidazol-1-yl)methyl]-2,3-dihydro-1H-carbazol-4-one;dihydrate
SMILES CC1=NC=CN1CC2CCC3=C(C2=O)C4=CC=CC=C4N3C.O.O.[Cl]

Specifications

Melting Point 178.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 365.85
Percent Purity 98%
Chemical Name or Material Ondansetron hydrochloride dihydrate
compliance-icons compliance-icons
Product Identifier
  • Ondansetron hydrochloride dihydrate, 98%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contains : Ondansetron hydrochloride dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ondansetron hydrochloride dihydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.