missing translation for 'onlineSavingsMsg'
Learn More

Köttermann One Door Tall Storage Cabinet

Product Code. p-7236282
Click to view available options
Hinge Side:
Left
Right
No. of Shelves:
3
4
Dimensions (D x W x H):
366 x 600 x 1920 mm
516 x 600 x 1920 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17302089

Brand: Köttermann 30100039

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

This One Door Tall Storage Cabinet features a single door to protect stored equipment and utensils.

  • Tall, slender design expands storage with a small footprint
  • Single door protects stored items

Specifications

Color White
Adjustable No
Cabinet Type Tall Storage
Dimensions (D x W x H) 366 x 600 x 1920 mm
Door Type Hinged
Finish Epoxy Coated
For Use With (Application) Storage
Hinge Side Left
Locking Mechanism Yes
No. of Doors 1
No. of Drawers None
No. of Shelves 4
Warranty None
Weight (Metric) 61.9 kg
With Casters No
Material Steel
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.