missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ LiquiSampler

Buerkle™ LiquiSampler for liquids, according to DIN EN ISO 15528. Made of ultra pure and chemically inert PTFE/FEP or PP for contamination-free sampling. Sampling from open and closed containers, barrels, tanks, silos, water courses. Suitable for all openings of at least 32 mm in diameter.

Brand:  Buerkle™ 5330-1100

274.88 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10115360

  • 321.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Practical one-handed use at the push of a button
  • With scraping ring for marking of depth and cleaning
  • Transparent
  • Available in PTFE/FEP or PP transparent
  • Point sampling, all-layer sampling and bottom sampling possible
  • Suitable for all commercially available barrels and containers with opening of at least 32mm in diameter
  • Easy to clean
Specifications

Specifications

LiquiSampler
Scraping ring
250 mL
DIN EN ISO 15528
32 mm
Polypropylene
Button - one-handed use
Transparent
Suitable for Use with Foodstuffs
100 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.