Learn More
Hamilton™ One-Hub Assembled Tubing Assemblies
Providing a variety of connection solutions in the laboratory.
Brand:  Hamilton™ 90613
 View more versions of this product
Product Code. 10170883
  • 40.00 € / Each
Explore more special offers
Description
Description
  • Fitting manufactured from CTFE
  • Tubing manufactured from PTFE
  • For female luers
  • A variety of gauges and lengths available
Specifications
Specifications
Tubing
PTFE
304.8 mm
13 G