Learn More
Hamilton™ One-Hub Assembled Tubing Assemblies
Providing a variety of connection solutions in the laboratory.
Brand: Hamilton™ 90666
This item is not returnable.
View return policy
Description
- Fitting manufactured from CTFE
- Tubing manufactured from PTFE
- For female luers
- A variety of gauges and lengths available
Specifications
Tubing Assembly | |
1/Pk. | |
16 ga. Connections | |
16 G |
Female Luer | |
CTFE (Fitting), PTFE (Tubing) | |
Custom |