Learn More
Hamilton™ One-Hub Assembled Tubing Assemblies
Providing a variety of connection solutions in the laboratory.
Brand:  Hamilton™ 90614
 View more versions of this product
Product Code. 11563084
  • 51.00 € / Each
Explore more special offers
Description
Description
  • Fitting manufactured from CTFE
  • Tubing manufactured from PTFE
  • For female luers
  • A variety of gauges and lengths available
Specifications
Specifications
Tubing Assembly
1/Pk.
14 ga. Connections
14 G
Female Luer
CTFE (Fitting), PTFE (Tubing)
304.8 mm