missing translation for 'onlineSavingsMsg'
Learn More

Ventum™ One-Touch™ Kit

Product Code. 17426893
Click to view available options
For Use With (Equipment):
Ventum One-Touch Pouches
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17426893

Brand: Ventum™ VAL 101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The One-Touch™ Kit contains a compressor, capsule, and accessories to use Ventum™ One-Touch™ Pouches.

  • Contains a compressor, a capsule, and a spray gun
  • Includes a wall and vehicle charger
  • Designed for use with Ventum™ One-Touch™ pouches

Specifications

For Use With (Equipment) Ventum One-Touch Pouches
Includes Compressor, capsule, and accessories to use Ventum™ One-Touch™ pouches
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.