Learn More
ONO-7300243, MedChemExpress
MedChemExpress ONO-7300243 is a novel, potent lysophosphatidic acid receptor 1 (LPA1) antagonist with IC50 of 0.16 μM.
Brand:  Medchem Express HY-100882
Encompass_Preferred
 View more versions of this product
Product Code. 16459756
  • 930.15 € / 50mg
Explore more special offers
Description
Description
ONO-7300243 is a novel, potent lysophosphatidic acid receptor 1 (LPA1) antagonist with IC50 of 0.16 μM.
Specifications
Specifications
ONO-7300243
White
C28H31NO5
DMSO : 100 mg/mL (216.66 mM; Need ultrasonic)
461.55
98.14%
Solid
638132-34-0
Research
50 mg
O=C(O)CC1=CC=C(CN(C(C2=CC(OC)=C(C)C(OC)=C2)=O)CCCC3=CC=CC=C3)C=C1
461.55
Research
COVID-19-immunoregulation