Learn More
Kolmi™ Op Air Pro Oxygen FFP3 NR D Mass Balance Masks
Op Air Pro Oxygen FFP3 masks are easy to put on with high breathability due to high quality raw material. Made with the Mass Balance approach from recycled materials.
Brand: Kolmi™ M53214-WH-MB
28.55 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Filtering half mask, efficiency class FFP3 NR D developed to protect against solid and liquid aerosols and particles
- Type IIR certified, it protects the environment against droplets emitted by the wearer and it protects from potential body fluid splashes
- Knitted headloops ultrasonically welded for optimal comfort
- Fits perfectly to face to limit inward leakage thanks to its shape and soft materials
- Hypoallergenic and has been dermatologically tested
- Made in France
Specifications
EN 149: 2001 + A1: 2009, DM Class I, EN 14683: 2019 + AC: 2019 | |
Small | |
Industry | |
Pouch |