missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Open Air Rockers

Ideal for cell culture work and are designed to be used in a variety of environmental conditions

925.00 €


Products 1
Product Code Brand Plug Type Certifications/Compliance Price Quantity & Availability  
Product Code Brand Plug Type Certifications/Compliance Price Quantity & Availability  
15514070
View accessory products View Documents
Fisherbrand™
88861026
EU, UK, CN Plug CE
925.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Low profile design: Takes up less space and easily fits into most fume hoods and incubators. Plastic base offers durability.
  • Microprocessor control: The microprocessor control provides electronic tilt angle adjustment which allows user to easily adjust rocking angle from 1 to 15° while unit is operating. Precise speed control provides smooth, low-speed rocking motion down to 1 rpm.
  • Memory: Display will show last user settings, even after power has been turned off. 
  • Continue to run by last set speed and the remaining time when the power recovered.
  • LED display: Touch pad controls with easy-to-read, LED displays for speed, tilt angle, and time . Provides repeatable and accurate results every time and is easily visible across lab benches. Timer will display elapsed time or, when programmed to user defined limit, will shut off unit when time reaches zero. Display will show last used settings, even after power has been shut off.
  • Applications: Staining and destaining gels, hybridization procedures, hematology and blotting technique

Warranty

2 years

Videos
SDS
Documents

Documents

Product Certifications