Learn More
Berkshire™ Open-Cell Foam Lab-Tips™ Swabs
General-purpose swab for cleaning small and confined areas
Brand:  Berkshire™ LTO70P.20
 View more versions of this product
Product Code. 17825510
  • 546.34 € / Pack of 20
Explore more special offers
Description
Description
  • Polyurethane foam over rigid core
  • 100 ppi open-cell foam
  • Thermally bonded head
  • Low particles, fibers, ions and extractables, no contaminating adhesives
  • Good chemical resistance
  • Good sorbency and excellent solvent hold
  • Laundered and packaged in cleanroom
Specifications
Specifications
ISO Class 4
2.8 in.
Swab
71 mm
5mm
Pointed
0.78 in.
3mm
0.098 in.
Designed for the highest level of contamination control in critical processing applications, applying lubricants and other liquids, removing excess materials
2.5mm
1.97 in.
Polyurethane Foam
3.8mm
2mm
1 in.