missing translation for 'onlineSavingsMsg'
Learn More

JULABO™ CORIO™ CD-BT5 Heating Circulator with Open Bath
BIOPHARMA

Julabo™ CORIO™ CD-BT5 Heating Circulator with Open Bath feature high quality pump connections and stainless steel bath tanks.

Brand:  JULABO™ 9012305

Product Code. 15410572

  • 1775.00 € / Each
Estimated Shipment: 03-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Model for internal and external applications
  • Bright, white, easy-to-read display
  • Very quiet
  • Easy pump change-over between internal and external circulation
  • USB connection
  • Class III (FL) according to DIN 12876-3

Applications:
Ideal for temperature control of samples, preparation of samples for serology, and clinical chemistry, analysis, material testing, and more. External temperature control applications in combination with measuring cells, photometers, refractometers, polarimeters, and more.

Specifications

Specifications

5 to 40 °C
20°C to 100°C
LED
0.35 bar
±0.03°C
38 x 23 x 38 cm
15 L/min.
Heating Circulator with Open Bath
2 kW
CORIO CD-BT5
15 x 15 x 15 cm
5.5 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers