missing translation for 'onlineSavingsMsg'
Learn More

Köttermann Open Tall Storage Cabinet

Product Code. p-7236284
Click to view available options
No. of Drawers:
4
None
No. of Shelves:
4
None
Dimensions (D x W x H):
350 x 600 x 1920 mm
350 x 900 x 1920 mm
500 x 600 x 1920 mm
500 x 900 x 1920 mm
350 x 1200 x 1920 mm
500 x 1200 x 1920 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17392079

Brand: Köttermann 30100033

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

The open-front design of this Open Tall Storage Cabinet provides easy access to stored equipment and utensils.

  • Tall, slender design expands storage with a small footprint
  • Open front provides easy access to stored items

Specifications

Color White
Adjustable No
Cabinet Type Tall Storage
Dimensions (D x W x H) 350 x 600 x 1920 mm
Door Type None
Finish Epoxy Coated
For Use With (Application) Storage
No. of Doors None
No. of Drawers None
No. of Shelves 4
Warranty None
Weight (Metric) 46.4 kg
With Casters No
Material Steel
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.