Learn More
Manutan Open top keg, 30 litre capacity
Brand:  Manutan A350763
Encompass_Preferred
Product Code. 12834108
  • 87.85 € / Each
Explore more special offers
Description
Description
Full open top, heavy duty kegs, made from polyethylene. Excellent resistance to most chemicals. Removable black lid retained by metal bond and tension cap. Ideal for food storage. UN Approved.
  • Capacity: 30L
  • Dimensions (H x D) : 520mm x 313mm
  • UN specification : 1H2/X 61/S