missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Open-Top Screw Caps

Thermo Scientific™ Open-Top Screw Caps provide an inert, airtight seal and direct puncture-access to sample with a syringe needle.

Brand:  Thermo Scientific™ TS-13218

 View more versions of this product

Product Code. 11501034

  • 58.47 € / Pack of 72
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Open-Top Screw Caps provide an inert, airtight seal and direct puncture-access to sample with a syringe needle.

  • Fit into Reacti-Vial™ small reaction vials
  • Used with PTFE/Rubber laminated discs
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fits 3 and 5mL Reacti-Vials, 40mL Screw Cap Septum Vials
Open Top Screw Cap
3 and 5 mL Reacti-Vials
3 mL
20 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.