missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ OpenArray™ 384-well Sample Plates, Black

For use with the QuantStudio™ 12K Flex Real-Time PCR System.

Brand:  Applied Biosystems™ 4482221

Product Code. 15826481

  • 126.00 € / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

They are low-volume microplates used in the QuantStudio™ OpenArray™ AccuFill™ System for sample loading onto OpenArray™ plates. These particular plates are black in color for improved visualization during loading.

4406947

Color: Clear, Reaction Speed: Standard 

NC0973054

Color: Black, Reaction Speed: Standard 
TRUSTED_SUSTAINABILITY
Specifications

Specifications

384-well Plate
QuantStudio™
OpenArray™
Real Time PCR (qPCR)
No Barcode
Each box contains 10 x 384-well plates, enough for 80 OpenArray™ Real-Time PCR plates.
Standard
Sample Plate
10 plates
Black
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.