Learn More
Hirschmann™ openend™ Disposable Capillary Pipettes, Na-hep
Excellent for working with small samples in the laboratory. Hirschmann Instruments™ openend™ Disposable Capillary Pipettes can be utilized for a wide range of scientific and medical applications.
Brand:  Hirschmann™ 905022001
Product Code. 10329901
  • 172.00 € / Pack of 1000
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Sodium Heparinized
  • Conformity certified
  • Manufactured from durable DURAN™ glass
  • CE acc. IVD 98/79 EC
Specifications
Specifications
0.5%
Glass
60 ±0.5 mm
Single-use
CE, IVD 98/79 EC
20 μL
Capillary Pipette