Learn More
Hirschmann™ Operating Cable
Operating cable for starting the dispensing process externally.
Brand:  Hirschmann™ 9564003
Product Code. 10319630
  • 42.15 € / Each
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For opus™
  • Connection cable between module and base unit
Specifications
Specifications
Starting the Dosing Operation Externally (for Basic Units 10 + 20 + 50 mL), e.g. For the Integration in Automatic Processes
1 Each
Control Cable