missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPRM1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | OPRM1 |
---|---|
Concentration | 267 μg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16885735
|
Proteintech
27625-1-AP-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16875735
|
Proteintech
27625-1-AP-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The OPRM1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human OPRM1. This antibody recognizes human, mouse, rat antigen. The OPRM1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
OPRM1 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
267 μg/mL | |
Polyclonal | |
Rabbit | |
4988 | |
Primary |