missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Opti-MEM™ I Reduced Serum Medium, GlutaMAX™ Supplement
Opti-MEM™ I Reduced Serum Medium, GlutaMAX™ Supplement
Brand: Gibco™ 51985026
This item is not returnable.
View return policy
Description
Opti-MEM™ Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change to growth rate or morphology. Opti-MEM™ is also recommended for use with cationic lipid transfection reagents, such as Lipofectamine™ reagent. Opti-MEM™ can be used with a variety of suspension and adherent mammalian cells, including Sp2, AE-1, CHO, BHK-21, HEK, and primary fibroblasts. We offer a variety of Gibco™ Opti-MEM™ modifications for a range of cell culture applications. The complete formulation is confidential. For more information, please contact Technical Services.Gibco™ Opti-MEM™ is a unique media that contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. These additional components allow for a reduction in serum supplementation by at least 50%. Gibco™ Opti-MEM™ with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ Opti-MEM™ is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ Opti-MEM™ product made in our Grand Island facility (31985-070). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Gibco™ Opti-MEM™ uses a sodium bicarbonate buffer system (2.4 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
ISO 13485 standards, FDA | |
Opti-Mem™ Reduced-Serum Medium, 500mL, Liquid, 1 X Concentration, 7.0 to 7.4 pH, With Glutamax™, Phenol Red, Sodium Pyruvate, 18 Months Shelf Life, 280 to 320mOsm/kg Osmolality, For Mammalian Cell Culture, Transfection | |
Liquid | |
GlutaMAX, Phenol Red, Sodium Pyruvate | |
7.0 to 7.4 |
2 to 8°C | |
1 X | |
Reduced-Serum Medium | |
Mammalian cell culture, transfection | |
500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For In Vitro Diagnostic Use.
Spot an opportunity for improvement?Share a Content Correction