Learn More
Thermo Scientific™ Opti-Bind™ Polystyrene Sulfate Particles
Achieve maximum reactivity in many diagnostic applications with Thermo Scientific™ Opti-Bind™ Polystyrene Sulfate Particles.
Brand: Thermo Scientific™ 81000597100290
Description
Achieve maximum reactivity in many diagnostic applications with Thermo Scientific™ Opti-Bind™ Polystyrene Sulfate Particles for use in turbidimetric/nephelometric assays, slide agglutination assays, chemiluminescent assays and in clinical diagnostics and molecular biology applications. Particles can be used direct from the bottle, without any pre-cleaning.
Our production does not utilize common surfactants (like SDS, Tween™ 20, Triton™ X-100) that can interfere with protein binding to particle surfaces. Polystyrene surfaces are very hydrophobic and adsorb proteins almost instantaneously. Our proprietary anionic surfactant does not interfere with the binding of proteins nor cause proteins to desorb from microparticle surfaces.- Available in a wide range of diameters (from 0.1 μM to 2.5 μM)
- No pre-washing
- Tight size distribution—CV<2%
- Optimized colloidal stability
- Shelf-life of 10 years
Recommended for:
Clinical diagnostics, molecular biology, nephelometric, turbidimetric and slide agglutination assays
Note:
When using Opti-Bind particles, ask about our Microparticle Reagent Optimization manual, which outlines our reagent development strategy. This manual includes validated binding, coupling and particle reagent optimization protocols.

Specifications
Includes Certificate of Analysis | |
Opti-Bind Polystyrene Sulfate Particles | |
Polystyrene Sulfate | |
Package Insert and Certificate of Analysis | |
These evaluation packs allow you to buy small samples from among our standard inventory of different surfaces, colors, sizes and binding capacities at greatly reduced prices. Opti-Bind particles are packaged in 15mL bottles at 10% solids. The evaluation pack consists of three lots from all available Opti-Bind polystyrene particles. | |
0.2 μm |
2°C to 8°C | |
Diagnostic Assays | |
Specialty Particles | |
15 mL | |
Opti-Bind Polystyrene particles are available in different concentrations and package sizes upon request. |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- 15ML OPTIBIND POLYSTYRENE PARTICLES, 0,2µM, 10% so lids, pasteurised, used in slide agglutination, lateral flow, and turbidimetric assays, Optibind microparticles are optimised for maximum reactivit y in many diagnostic applications 15mL Ship at
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
Hazard Statement
- H351-Suspected of causing cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : polstyrene, sodium azide
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
274531_STD.STK_1199 | Certificate of Analysis | 28/01/2025 | 81000597100290 |
278534_STD.STK_1199 | Certificate of Analysis | 26/01/2025 | 81000597100290 |
281801_STD.STK_1199 | Certificate of Analysis | 26/01/2025 | 81000597100290 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.