Learn More
Hellma™ Special Optical Glass Precision Micro Cell Cuvettes
Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Special Optical Glass Precision Micro Cell Cuvettes are manufactured from SCHOTT™ type UK 5 glass, which is made from exceptionally pure raw materials, which gives an improved transmission in the near ultraviolet range.
Brand:  Hellma™ 100-100-20
 View more versions of this product
Product Code. 10437641
  • 318.00 € / Each
Estimated Shipment: 23-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Transmission of more than 80% over a spectral range of between 320 and 2500nm
  • For photometry and colorimetry measurements
Specifications
Specifications
Macro Absorption Cell
Optical Glass
100 mm
45 mm
35,000 μL
9.5mm
360 to 2500 nm
12.5 mm