Learn More
Hellma™ Optical Glass Micro Cells
Brand:  Hellma™ 6030-50-10
 View more versions of this product
Product Code. 10673711
  • 151.00 € / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Micro Cell
Optical Glass
9.5mm
50 mm
45 mm
17.5 mL
PTFE
1Each
360 to 2500 nm
12.5 mm