missing translation for 'onlineSavingsMsg'
Learn More

Biochrom Optical Glass Standard Cuvette with Lid

Optical Glass Standard Cuvette features standard rectangular cell with lid for visible/UV spectrophotometry.

Brand:  Biochrom 80-2003-87

 View more versions of this product

Product Code. 11310142

  • 121.00 € / Each
Estimated Shipment: 16-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Optical Glass
  • Standard rectangular cell with lid
  • Fits standard spectrophotometer cell holders
Specifications

Specifications

Type 0 Standard Cuvette
Optical Glass
10 mm
10 mm
45 mm
45 mm
Rectangle
2 mL
12.5 mm
1 Ea.
190 to 1100 nm
Spectrophotometer, Photometer
Spectrophotometric Measurements
12.7 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers