missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Optifit Tips, Refills
GREENER_CHOICE

Product Code. 15161037 Shop All Sartorius Products
Click to view available options
Color Code:
Blue
Gray
Green
Orange
Purple
Red
Yellow
Sterility:
Non-sterile
Sterile
Volume (Metric):
0.1 to 10 μL
0.5 to 300 μL
10 to 1000 μL
100 to 5000 μL
1000 to 10,000 μL
5 to 350 μL
50 to 1200 μL
Packaging:
Bulk in Box
Bulk in Carton
Refill Pack
Refill Tower
Refill Twin Pack
Unit Size:
Pack of 1000
Pack of 250
Pack of 960
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 15161037

Brand: Sartorius LH780316

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Ensure perfect fitting and leak-tight sealing on Sartorius pipettes. Sartorius Optifit Tips, Refills promote maximum pipetting accuracy and reproducibility. Tips are manufactured in an ISO 8 certified cleanroom environment and are available in sterile and non-sterile versions in a variety of volumes.

Ergonomically designed for maximum precision and accuracy
  • Perfect tip fitting and sealing to the pipetter
  • Enables ergonomic and light tip attachment and ejection
  • Precision made tips provide best possible accuracy and precision
Compatible with Optiload feature in Sartorius pipetters and more:
  • Picus, eLINE™, mLINE™ and Proline™ Plus
  • Color-coded trays and tips to easily match with compatible Sartorius pipetters
  • Universal design is compatible with most other brands of pipetters
Designed to the highest standards
  • Made from high-quality virgin polypropylene
  • Refill Packs are DNase, RNase and endotoxin-free certified
  • Available as e-beam pre-sterilized or non-sterilized
  • Improved tip identification and traceability by informative rack labels
    • Indicates tip volume, product number, lot number and purity level
  • Packaging and tips are 100% recyclable or can be incinerated
  • Available in a range of packaging formats:
    • Refill Packs - purity certified and ecological packages
    • Refill Towers - space-saving and ecological
    • Bulk Cartons - economical bulk boxes
  • Air-tight plastic wrapping around the rack secures purity during transport and storage
  • Reusable and fully autoclavable at 121°C for 20 minutes

Certifications

CE certified. Manufactured according to ISO 9001 and ISO 13485.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1000 to 10,000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Transparent
Autoclavable Autoclavable
Length (Metric) 15.5cm
Certifications/Compliance CE, ISO 9001, ISO 13485
Sterilization Method Autoclave at 121°C
Format Bulk, Bagged
Tip Style Standard
Packaging Bulk in Box
Product Type Non-filtered Tips
Color Code Red
Disposable Single-use
For Use With (Equipment) Sartorius Proline Plus, Sartorius mLINE, Sartorius eLINE, Sartorius Picus
Graduated No
Product Line Optifit
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.